Dimethylpiperidin-2-ylmethylamine dihydrochloride 97%
Dodavatel: Apollo Scientific
Synonyms:
2-(Dimethylaminomethyl)piperidine dihydrochloride
OR452113-1G
OR452113-250MG
OR452113-500MG
APOSOR452113-1GEA
18620
CZK
APOSOR452113-1G
APOSOR452113-250MG
APOSOR452113-500MG
Dimethylpiperidin-2-ylmethylamine dihydrochloride 97%
Dimethylpiperidin-2-ylmethylamine dihydrochloride
Vzorec:
C₈H₂₀Cl₂N₂ MW: 215,17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09879171 CAS číslo: 1185304-33-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...