Tisk…

1-Iodo-4-phenoxybenzene 97%

Dodavatel: Apollo Scientific

OR452072-5G
APOSOR452072-5GEA 1660 CZK
APOSOR452072-5G
1-Iodo-4-phenoxybenzene 97%
1-Iodo-4-phenoxybenzene
Vzorec: C₁₂H₉IO
MW: 296,1037 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00060666
CAS číslo: 2974-94-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR