2-Methylphenethyl bromide 95%
Dodavatel: Apollo Scientific
OR452071-1G
OR452071-5G
APOSOR452071-1GEA
0
CZK
APOSOR452071-1G
APOSOR452071-5G
2-Methylphenethyl bromide 95%
2-Methylphenethyl bromide
Vzorec:
C₉H₁₁Br Storage Temperature: Ambient |
MDL Number:
MFCD04117467 CAS číslo: 16793-90-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...