Tisk…

3-Methoxyphenethyl bromide 97%

Dodavatel: Apollo Scientific

Synonyms: 1-(2-Bromoethyl)-3-methoxybenzene

OR452070-1G OR452070-5G
APOSOR452070-1GEA 2150 CZK
APOSOR452070-1G APOSOR452070-5G
3-Methoxyphenethyl bromide 97%
3-Methoxyphenethyl bromide
Vzorec: C₉H₁₁BrO
Storage Temperature: Ambient
MDL Number: MFCD07369789
CAS číslo: 2146-61-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR