Tisk…

((4-Iodophenyl)ethynyl)trimethylsilane 97%

Dodavatel: Apollo Scientific

Synonyms: 1-Iodo-4-[2-(trimethylsilyl)ethynyl]-benzene

OR452068-1G OR452068-5G
APOSOR452068-1GEA 1700 CZK
APOSOR452068-1G APOSOR452068-5G
((4-Iodophenyl)ethynyl)trimethylsilane 97%
((4-Iodophenyl)ethynyl)trimethylsilane
Vzorec: C₁₁H₁₃ISi
MW: 300,214 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04974026
CAS číslo: 134856-58-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR