Tisk…

4-(3-Chlorophenoxy)benzoic acid 97%

Dodavatel: Apollo Scientific

OR452063-1G OR452063-250MG
APOSOR452063-1GEA 14320 CZK
APOSOR452063-1G APOSOR452063-250MG
4-(3-Chlorophenoxy)benzoic acid 97%
4-(3-Chlorophenoxy)benzoic acid
Vzorec: C₁₃H₉ClO₃
Storage Temperature: Ambient
MDL Number: MFCD11122181
CAS číslo: 1145-58-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR