Boc-7-methyl-DL-tryptophan 97%
Dodavatel: Apollo Scientific
OR452045-1G
OR452045-250MG
OR452045-500MG
APOSOR452045-1GEA
27910
CZK
APOSOR452045-1G
APOSOR452045-250MG
APOSOR452045-500MG
Boc-7-methyl-DL-tryptophan 97%
Boc-7-methyl-DL-tryptophan
Vzorec:
C₁₇H₂₂N₂O₄ Storage Temperature: Ambient |
MDL Number:
MFCD02682378 CAS číslo: 1219333-83-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...