Tisk…

Fmoc-DL-β-leucine 97%

Dodavatel: Apollo Scientific

Synonyms: 3-(Fmoc-amino)-4-methylpentanoic acid

OR452042-1G OR452042-500MG
APOSOR452042-1GEA 16770 CZK
APOSOR452042-1G APOSOR452042-500MG
Fmoc-DL-β-leucine 97%
Fmoc-DL-beta-leucine
Vzorec: C₂₁H₂₃NO₄
MW: 353.42 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01863209
CAS číslo: 282524-93-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR