Boc-DL-β-leucine 97%
Dodavatel: Apollo Scientific
Synonyms:
3-(Boc-amino)-4-methylpentanoic acid, DL-3-((tert-Butoxycarbonyl)amino)-4-methylpentanoic acid, Boc-DL-β-HoVal-OH
OR452041-1G
APOSOR452041-1GEA
2240
CZK
APOSOR452041-1G
Boc-DL-β-leucine 97%
Boc-DL-beta-leucine
Vzorec:
C₁₁H₂₁NO₄ MW: 231,29 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02258127 CAS číslo: 248924-39-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...