Tisk…

(1-Benzothien-2-ylmethyl)methylamine hydrochloride

Dodavatel: Apollo Scientific
Varování

OR451442-1G OR451442-5G
APOSOR451442-1GEA 1910 CZK
APOSOR451442-1G APOSOR451442-5G
(1-Benzothien-2-ylmethyl)methylamine hydrochloride
(1-Benzothien-2-ylmethyl)methylamine hydrochloride
Vzorec: C₁₀H₁₂ClNS
MW: 213,73 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03306023
CAS číslo: 849776-43-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR