Tisk…

(3-Nitro-1H-pyrazol-1-yl)acetonitrile

Dodavatel: Apollo Scientific
Varování

OR451438-1G OR451438-5G
APOSOR451438-1GEA 3110 CZK
APOSOR451438-1G APOSOR451438-5G
(3-Nitro-1H-pyrazol-1-yl)acetonitrile
(3-Nitro-1H-pyrazol-1-yl)acetonitrile
Vzorec: C₅H₄N₄O₂
Storage Temperature: Ambient
MDL Number: MFCD04969694
CAS číslo: 1006956-03-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR