5-Amino-4-chloro-1H-pyrazole-3-carboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR451434-1G
OR451434-25G
OR451434-5G
APOSOR451434-1GEA
744
CZK
APOSOR451434-1G
APOSOR451434-25G
APOSOR451434-5G
5-Amino-4-chloro-1H-pyrazole-3-carboxylic acid
5-Amino-4-chloro-1H-pyrazole-3-carboxylic acid
Vzorec:
C₄H₄ClN₃O₂ Storage Temperature: Ambient |
MDL Number:
MFCD15146532 CAS číslo: 351990-69-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...