Tisk…

Methyl-1-(chloromethyl)-1H-pyrazole-5-carboxylate

Dodavatel: Apollo Scientific
Nebezpečí

OR451428-1G OR451428-25G OR451428-5G
APOSOR451428-1GEA 7440 CZK
APOSOR451428-1G APOSOR451428-25G APOSOR451428-5G
Methyl-1-(chloromethyl)-1H-pyrazole-5-carboxylate
Methyl-1-(chloromethyl)-1H-pyrazole-5-carboxylate
Vzorec: C₆H₇ClN₂O₂
MW: 174,59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04969151
CAS číslo: 1001499-93-9
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR