Tisk…

1-(Chloromethyl)-3,5-dimethyl-4-nitro-1H-pyrazole

Dodavatel: Apollo Scientific
Nebezpečí

OR451427-1G OR451427-5G
APOSOR451427-1GEA 1370 CZK
APOSOR451427-1G APOSOR451427-5G
1-(Chloromethyl)-3,5-dimethyl-4-nitro-1H-pyrazole
1-(Chloromethyl)-3,5-dimethyl-4-nitro-1H-pyrazole
Vzorec: C₆H₈ClN₃O₂
Storage Temperature: Ambient
MDL Number: MFCD04968629
CAS číslo: 51355-91-0
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR