Tisk…

3-Methoxy-5-nitroaniline

Dodavatel: Apollo Scientific
Varování

OR451426-1G OR451426-5G
APOSOR451426-1GEA 3310 CZK
APOSOR451426-1G APOSOR451426-5G
3-Methoxy-5-nitroaniline
3-Methoxy-5-nitroaniline
Vzorec: C₇H₈N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD01103799
CAS číslo: 586-10-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR