2,2'-Dithienyl disulphide 95+%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1,2-Di(thiophen-2-yl)disulphane, Di(2-thienyl) Disulphide
OR451331-1G
OR451331-25G
OR451331-5G
APOSOR451331-1GEA
622
CZK
APOSOR451331-1G
APOSOR451331-25G
APOSOR451331-5G
2,2'-Dithienyl disulphide 95+%
2,2'-Dithienyl disulphide
Vzorec:
C₈H₆S₄ MW: 230,38 g/mol Bod varu: 132 °C Bod tání: 58 °C Storage Temperature: Ambient |
MDL Number:
MFCD00066333 CAS číslo: 6911-51-9 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...