Diphenyliodonium chloride
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR451320-25G
OR451320-5G
OR451320-100G
APOSOR451320-25GEA
3480
CZK
APOSOR451320-25G
APOSOR451320-5G
APOSOR451320-100G
Diphenyliodonium chloride
Diphenyliodonium chloride
Vzorec:
C₁₂H₁₀ClI MW: 316,57 g/mol Bod tání: 233…235 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011909 CAS číslo: 1483-72-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...