1-(Cyanomethyl)pyridinium chloride 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-(Cyanomethyl)pyridin-1-ium chloride
OR451315-100G
OR451315-25G
APOSOR451315-100GEA
10190
CZK
APOSOR451315-100G
APOSOR451315-25G
1-(Cyanomethyl)pyridinium chloride 97%
1-(Cyanomethyl)pyridinium chloride
Vzorec:
C₇H₇ClN₂ MW: 154,60 g/mol Bod tání: 169…174 °C Storage Temperature: Ambient |
MDL Number:
MFCD00031933 CAS číslo: 17281-59-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...