6-Phenyl-1-hexanol 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
6-Phenylhexanol
OR451311-10G
OR451311-1G
OR451311-5G
APOSOR451311-10GEA
9860
CZK
APOSOR451311-10G
APOSOR451311-1G
APOSOR451311-5G
6-Phenyl-1-hexanol 98%
6-Phenyl-1-hexanol
Vzorec:
C₁₂H₁₈O MW: 178,27 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00014063 CAS číslo: 2430-16-2 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...