3-Benzylidene-2,4-pentanedione 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR451308-25G
OR451308-5G
APOSOR451308-25GEA
5130
CZK
APOSOR451308-25G
APOSOR451308-5G
3-Benzylidene-2,4-pentanedione 97%
3-Benzylidene-2,4-pentanedione
Vzorec:
C₁₂H₁₂O₂ MW: 188,23 g/mol Bod varu: 185…188 °C Hustota: 1,081 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00014990 CAS číslo: 4335-90-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...