N,N-Dimethylglycine 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-(Dimethylamino)acetic acid, (Dimethylamino)acetic acid
OR45130-25G
OR45130-5G
OR45130-100G
APOSOR45130-25GEA
827
CZK
APOSOR45130-25G
APOSOR45130-5G
APOSOR45130-100G
N,N-Dimethylglycine 97%
N,N-Dimethylglycine
Vzorec:
C₄H₉NO₂ MW: 103,12 g/mol Bod tání: 178…183 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004283 CAS číslo: 1118-68-9 EINECS: 214-267-8 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...