Tisk…

3-Bromo-p-anisidine 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 3-Bromo-4-methoxyaniline

OR45122-1G OR45122-5G
APOSOR45122-1GEA 622 CZK
APOSOR45122-1G APOSOR45122-5G
3-Bromo-p-anisidine 97%
3-Bromo-p-anisidine
Vzorec: C₇H₈BrNO
MW: 202,0485 g/mol
Bod tání: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD04094339
CAS číslo: 19056-41-8
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR