Tisk…

(3,3-Dimethoxycyclobutyl)methanol

Dodavatel: Apollo Scientific

OR451123-1G OR451123-10MG OR451123-250MG OR451123-500MG OR451123-2.5G
APOSOR451123-1GEA 4860 CZK
APOSOR451123-1G APOSOR451123-10MG APOSOR451123-250MG APOSOR451123-500MG APOSOR451123-2.5G
(3,3-Dimethoxycyclobutyl)methanol
(3,3-Dimethoxycyclobutyl)methanol
Vzorec: C₇H₁₄O₃
MW: 146,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11110681
CAS číslo: 175021-11-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR