Tisk…

N-tert-Butylhydroxylamine

Dodavatel: Apollo Scientific

Synonyms: N-(tert-Butyl)hydroxylamine

OR45111-250MG
APOSOR45111-250MGEA 5200 CZK
APOSOR45111-250MG
N-tert-Butylhydroxylamine
N-tert-Butylhydroxylamine
Vzorec: C₄H₁₁NO
MW: 89,14 g/mol
Bod tání: 59…65 °C
Hustota: 0,88 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD06653532
CAS číslo: 16649-50-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR