Tisk…

(R)-1-Boc-2-Isopropylpiperazine

Dodavatel: Apollo Scientific
Varování

OR451095-1G OR451095-5G
APOSOR451095-1GEA 3440 CZK
APOSOR451095-1G APOSOR451095-5G
(R)-1-Boc-2-Isopropylpiperazine
(R)-1-Boc-2-Isopropylpiperazine
Vzorec: C₁₂H₂₄N₂O₂
MW: 228,3312 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07772101
CAS číslo: 674792-04-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR