Tisk…

(2-Mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)acetic acid

Dodavatel: Apollo Scientific

OR450085-1G OR450085-250MG
APOSOR450085-1GEA 4960 CZK
APOSOR450085-1G APOSOR450085-250MG
(2-Mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)acetic acid
(2-Mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)acetic acid
Vzorec: C₅H₅NO₃S₂
Storage Temperature: Ambient
MDL Number: MFCD12027984
CAS číslo: 701-23-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR