Tisk…

8-Bromo-6-methyl-3-nitroimidazo[1,2-a]pyridine

Dodavatel: Apollo Scientific

OR450078-1G OR450078-25G OR450078-5G
APOSOR450078-1GEA 2070 CZK
APOSOR450078-1G APOSOR450078-25G APOSOR450078-5G
8-Bromo-6-methyl-3-nitroimidazo[1,2-a]pyridine
8-Bromo-6-methyl-3-nitroimidazo[1,2-a]pyridine
Vzorec: C₈H₆BrN₃O₂
Storage Temperature: Ambient
MDL Number: MFCD11504901
CAS číslo: 1072944-59-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR