Tisk…
1,3-Bis(1-adamantyl)benzimidazolium chloride 97%
Dodavatel: Apollo Scientific
Synonyms: 1,3-Bis(1-adamanthyl)benzimidazolium chloride, 1,3-Di(adamantan-1-yl)-1H-benzo[d]imidazol-3-ium chloride
OR450037-1G OR450037-250MG
APOSOR450037-1GEA 9720 CZK
APOSOR450037-1G APOSOR450037-250MG
1,3-Bis(1-adamantyl)benzimidazolium chloride 97%
1,3-Bis(1-adamantyl)benzimidazolium chloride
Vzorec: C₂₇H₃₅N₂·Cl
MW: 423.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08705246
CAS číslo: 852634-41-4