Tisk…

2-Bromo-4-methylthiazole-5-carboxylic acid 95%

Dodavatel: Apollo Scientific

Synonyms: 2-Bromo-4-methyl-5-thiazolecarboxylic acid, 2-bromo-4-methyl-1,3-thiazole-5-carboxylic acid

OR450004-25G
APOSOR450004-25GEA 2900 CZK
APOSOR450004-25G
2-Bromo-4-methylthiazole-5-carboxylic acid 95%
2-Bromo-4-methylthiazole-5-carboxylic acid
Vzorec: C₅H₄BrNO₂S
MW: 222,06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09026784
CAS číslo: 40003-41-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR