Tisk…

5-Bromopyrazolo[1,5-a]pyrimidine 97%

Dodavatel: Apollo Scientific

OR450002-1G
APOSOR450002-1GEA 1620 CZK
APOSOR450002-1G
5-Bromopyrazolo[1,5-a]pyrimidine 97%
5-Bromopyrazolo[1,5-a]pyrimidine
Vzorec: C₆H₄BrN₃
MW: 198,02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11042744
CAS číslo: 1159981-95-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR