4-(Methylsulphonyl)aniline hydrochloride 95+%
Dodavatel: Apollo Scientific
|
Varování
|
OR4488-1G
OR4488-250MG
OR4488-5G
APOSOR4488-1GEA
705
CZK
APOSOR4488-1G
APOSOR4488-250MG
APOSOR4488-5G
4-(Methylsulphonyl)aniline hydrochloride 95+%
4-(Methylsulphonyl)aniline hydrochloride
Vzorec:
C₇H₁₀ClNO₂S MW: 207,67 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00216490 CAS číslo: 177662-76-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...