Tisk…

4-(3-Butyl-1-imidazolio)-1-butanesulphonate 95%

Dodavatel: Apollo Scientific
Varování

OR4470-1G OR4470-5G
APOSOR4470-1GEA 5290 CZK
APOSOR4470-1G APOSOR4470-5G
4-(3-Butyl-1-imidazolio)-1-butanesulphonate 95%
4-(3-Butyl-1-imidazolio)-1-butanesulfonate
Vzorec: C₁₁H₂₀N₂O₃S
Storage Temperature: Ambient
MDL Number: MFCD07784454
CAS číslo: 439937-61-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR