3-Carboxyphenylboronic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR4442-100G
OR4442-25G
OR4442-5G
APOSOR4442-100GEA
2610
CZK
APOSOR4442-100G
APOSOR4442-25G
APOSOR4442-5G
3-Carboxyphenylboronic acid
3-Carboxyphenylboronic acid
Vzorec:
C₇H₇BO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00036833 CAS číslo: 25487-66-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...