Tisk…

[4-(5-Methyl-1,2,4-oxadiazol-3-yl)phenyl]methanol 97%

Dodavatel: Apollo Scientific
Varování

OR4419-1G OR4419-5G OR4419-250MG
APOSOR4419-1GEA 7280 CZK
APOSOR4419-1G APOSOR4419-5G APOSOR4419-250MG
[4-(5-Methyl-1,2,4-oxadiazol-3-yl)phenyl]methanol 97%
[4-(5-Methyl-1,2,4-oxadiazol-3-yl)phenyl]methanol
Vzorec: C₁₀H₁₀N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD07368531
CAS číslo: 852180-61-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR