Tisk…

3-[3-(Bromomethyl)phenyl]-5-methyl-1,2,4-oxadiazole 95%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 3-(3-(Bromomethyl)phenyl)-5-methyl-1,2,4-oxadiazole

OR4411-1G OR4411-250MG
APOSOR4411-1GEA 29760 CZK
APOSOR4411-1G APOSOR4411-250MG
3-[3-(Bromomethyl)phenyl]-5-methyl-1,2,4-oxadiazole 95%
3-[3-(Bromomethyl)phenyl]-5-methyl-1,2,4-oxadiazole
Vzorec: C₁₀H₉BrN₂O
Bod tání: 71…72 °C
Storage Temperature: Ambient
MDL Number: MFCD07368547
CAS číslo: 253273-90-4
UN: 3261
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR