tert-Butyl-4-oxoazepane-1-carboxylate 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Tert-butyl 4-oxoazepane-1-carboxylate, 4-Oxoazepane-1-carboxylic acid tert-butyl ester, N-BOC-Hexahydro-1H-azepin-4-one, tert-Butyl 4-oxoazepane-1-carboxylate
OR4399-1G
APOSOR4399-1GEA
622
CZK
APOSOR4399-1G
tert-Butyl-4-oxoazepane-1-carboxylate 95%
tert-Butyl-4-oxoazepane-1-carboxylate
Vzorec:
C₁₁H₁₉NO₃ MW: 213,277 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD03788435 CAS číslo: 188975-88-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...