4-(2-Methylthiazol-4-yl)phenylamine 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-(2-Methylthiazol-4-yl)aniline, 4-(2-Methyl-1,3-thiazol-4-yl)aniline
OR4380-250MG
OR4380-5G
APOSOR4380-250MGEA
1040
CZK
APOSOR4380-250MG
APOSOR4380-5G
4-(2-Methylthiazol-4-yl)phenylamine 97%
4-(2-Methylthiazol-4-yl)phenylamine
Vzorec:
C₁₀H₁₀N₂S MW: 190,27 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD02691928 CAS číslo: 25021-49-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...