Tisk…

2-(1-Pyrrolidinyl)-5-pyrimidinecarboxylic acid 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-(pyrrolidin-1-yl)pyrimidine-5-carboxylic acid

OR43686-1G OR43686-250MG
APOSOR43686-1GEA 5390 CZK
APOSOR43686-1G APOSOR43686-250MG
2-(1-Pyrrolidinyl)-5-pyrimidinecarboxylic acid 97%
2-(1-Pyrrolidinyl)-5-pyrimidinecarboxylic acid
Vzorec: C₉H₁₁N₃O₂
MW: 193,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09054869
CAS číslo: 253315-06-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR