Tisk…

tert-Butyl furan-3-ylcarbamate 95+%

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Aminofuran, N-BOC protected

OR43662-5G
APOSOR43662-5GEA 2730 CZK
APOSOR43662-5G
tert-Butyl furan-3-ylcarbamate 95+%
tert-Butyl furan-3-ylcarbamate
Vzorec: C₉H₁₃NO₃
MW: 183,2044 g/mol
Bod tání: 135…140 °C
Storage Temperature: Ambient
MDL Number: MFCD09839085
CAS číslo: 56267-48-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR