4,6-Dichloro-7-azaindole 98%
Dodavatel: Apollo Scientific
|
Varování
|
OR43659-1G
OR43659-5G
APOSOR43659-1GEA
1620
CZK
APOSOR43659-1G
APOSOR43659-5G
4,6-Dichloro-7-azaindole 98%
4,6-Dichloro-7-azaindole
Vzorec:
C₇H₄Cl₂N₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD09880157 CAS číslo: 5912-18-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...