N-Methyl-N-(4-morpholin-4-ylbenzyl)amine 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR4360-1G
OR4360-250MG
APOSOR4360-1GEA
2150
CZK
APOSOR4360-1G
APOSOR4360-250MG
N-Methyl-N-(4-morpholin-4-ylbenzyl)amine 97%
N-Methyl-N-(4-morpholin-4-ylbenzyl)amine
Vzorec:
C₁₂H₁₈N₂O Storage Temperature: Ambient |
MDL Number:
MFCD07348565 CAS číslo: 179328-22-4 UN: 1759 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...