3-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-Methyl-1H-pyrazole-4-boronic acid, pinacol ester
OR43573-1G
OR43573-10G
APOSOR43573-1GEA
622
CZK
APOSOR43573-1G
APOSOR43573-10G
3-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 98%
3-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Vzorec:
C10H17BN2O2 MW: 208,0652 g/mol Bod tání: 129…134 °C Storage Temperature: Ambient |
MDL Number:
MFCD08690235 CAS číslo: 936250-20-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...