6-Bromoindazole-1-carboxylic acid tert-butyl ester
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
tert-Butyl 6-bromo-1H-indazole-1-carboxylate, 6-Bromo-1H-indazole, N1-BOC protected
OR43564-1G
OR43564-5G
OR43564-250MG
APOSOR43564-1GEA
1240
CZK
APOSOR43564-1G
APOSOR43564-5G
APOSOR43564-250MG
6-Bromoindazole-1-carboxylic acid tert-butyl ester
6-Bromoindazole-1-carboxylic acid tert-butyl ester
Vzorec:
C₁₂H₁₃BrN₂O₂ MW: 297,16 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD06739133 CAS číslo: 877264-77-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...