5-Chloropyrazine-2-carbonitrile 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR43502-1G
OR43502-5G
OR43502-10G
APOSOR43502-1GEA
910
CZK
APOSOR43502-1G
APOSOR43502-5G
APOSOR43502-10G
5-Chloropyrazine-2-carbonitrile 97%
5-Chloropyrazine-2-carbonitrile
Vzorec:
C5H2ClN3 Storage Temperature: Refrigerator |
MDL Number:
MFCD09607700 CAS číslo: 36070-75-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...