3-Bromo-5-iodobenzoic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR4328-25G
OR4328-5G
APOSOR4328-25GEA
1740
CZK
APOSOR4328-25G
APOSOR4328-5G
3-Bromo-5-iodobenzoic acid
3-Bromo-5-iodobenzoic acid
Vzorec:
C₇H₄BrIO₂ MW: 326,92 g/mol Bod tání: 219…221 °C Storage Temperature: Freezer |
MDL Number:
MFCD00191851 CAS číslo: 188815-32-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...