Tisk…

4-Chlorophenylhydrazinium chloride 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Chlorophenylhydrazine hydrochloride, 1-(4-Chlorophenyl)hydrazine hydrochloride

OR4295-500G
APOSOR4295-500GEA 2070 CZK
APOSOR4295-500G
4-Chlorophenylhydrazinium chloride 97%
4-Chlorophenylhydrazinium chloride
Vzorec: C₆H₈Cl₂N₂
MW: 179,05 g/mol
Bod tání: 216 °C (rozkl.)
Storage Temperature: Refrigerator
MDL Number: MFCD00012943
CAS číslo: 1073-70-7
EINECS: 214-030-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR