4-Amino-3-iodopyridine
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR4266-1G
OR4266-5G
APOSOR4266-1GEA
622
CZK
APOSOR4266-1G
APOSOR4266-5G
4-Amino-3-iodopyridine
4-Amino-3-iodopyridine
Vzorec:
C₅H₅IN₂ Bod tání: 95…101 °C Storage Temperature: Ambient |
MDL Number:
MFCD02663853 CAS číslo: 88511-27-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...