2-Chloro-5-(chloromethyl)thiazole
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR4254-25G
OR4254-5G
OR4254-100G
APOSOR4254-25GEA
744
CZK
APOSOR4254-25G
APOSOR4254-5G
APOSOR4254-100G
2-Chloro-5-(chloromethyl)thiazole
2-Chloro-5-(chloromethyl)thiazole
Vzorec:
C₄H₃Cl₂NS MW: 168,0443 g/mol Bod tání: 29…31 °C Hustota: 1,503 g/cm³ (25 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD01073549 CAS číslo: 105827-91-6 EINECS: 429-830-5 UN: 2922 ADR: 8+6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...