4-Nitrophenyl bromoacetate 98%
Dodavatel: Apollo Scientific
|
Varování
|
OR42285-1G
OR42285-5G
OR42285-10G
APOSOR42285-1GEA
1210
CZK
APOSOR42285-1G
APOSOR42285-5G
APOSOR42285-10G
4-Nitrophenyl bromoacetate 98%
4-Nitrophenyl bromoacetate
Vzorec:
C₈H₆BrNO₄ Bod tání: 83…85 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00051840 CAS číslo: 19199-82-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...