4,4-Dimethylcyclohexene-1-ylboronic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(4,4-Dimethylcyclohex-1-en-1-yl)boronic acid
OR4226-250MG
OR4226-1G
APOSOR4226-250MGEA
9100
CZK
APOSOR4226-250MG
APOSOR4226-1G
4,4-Dimethylcyclohexene-1-ylboronic acid 98%
4,4-Dimethylcyclohexene-1-ylboronic acid
Vzorec:
C₈H₁₅BO₂ Bod tání: 152…154 °C Storage Temperature: Freezer |
MDL Number:
MFCD07363758 CAS číslo: 865869-28-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...